Learn More
Genscript Corporation MonoRab™ Rabbit Anti-Camelid VHH Antibody [iFluor488], mAb
Supplier:  Genscript Corporation A01862200
Encompass
GenScript MonoRab™ Rabbit Anti-Camelid VHH Antibody [iFluor488], mAb is specific for Camelid VHH Antibody (Alpaca, Llama and Camel). The monoclonal antibody has no cross-reactivity with mouse, rat, rabbit, chicken, goat or human immunoglobulins.
Catalog No. NC1550740