Learn More
Genscript Corporation Antibody Antibody Antibodies Antibodies 200 UL 1/EA
Supplier:  Genscript Corporation A02227200
Encompass
The product is specific for Camelid (Llama Alpaca and Camel) heavy chain antibodies (IgG2and3) but does not recognize the camelid conventional antibody (IgG1).
Catalog No. 50-253-0019