Learn More
Genscript Corporation MonoRab™ Rabbit Anti-Camelid VHH Antibody, mAb
Catalog No. NC1480417
Encompass
GenScript MonoRab™ Rabbit Anti-Camelid VHH Antibody, mAb is specific for Camelid sdAb (Alpaca, Llama and Camel). The monoclonal antibody has no cross-reactivity with mouse, rat, rabbit, chicken, goat or human immunoglobulins.
Catalog No. NC1480417 Supplier Genscript Corporation Supplier No. A01860200
GenScript MonoRab™ Rabbit Anti-Camelid VHH Antibody, mAb is specific for Camelid sdAb (Alpaca, Llama and Camel). The monoclonal antibody has no cross-reactivity with mouse, rat, rabbit, chicken, goat or human immunoglobulins.