Learn More
Genscript Corporation MonoRab™ Rabbit Anti-Humanized VHH Antibody [Biotin], mAb
Supplier:  Genscript Corporation A02166200
Encompass
The product is specific for Humanized VHH antibodies and Camelid (Llama, Alpaca, and Camel) heavy chain antibodies (IgG2&3), but does not recognize the camelid conventional antibody (IgG1).
Catalog No. NC2137107