Learn More
Genscript Corporation Antibody Antibody Antibodies Antibodies 200 UL 1/EA
Supplier:  Genscript Corporation A02172200
Encompass
The product is specific for Humanized VHH antibodies and Camelid (Llama Alpaca and Camel) heavy chain antibodies (IgG2and3) but does not recognize the camelid conventional antibody (IgG1).
Catalog No. 50-253-0036