Learn More
Genscript Corporation Antibody Antibody Antibodies Antibodies 200 UL 1/EA
Catalog No. 502530038
Encompass
The product is specific for Humanized VHH antibodies and Camelid (Llama Alpaca and Camel) heavy chain antibodies (IgG2and3) but does not recognize the camelid conventional antibody (IgG1).
Catalog No. 50-253-0038 Supplier Genscript Corporation Supplier No. A02168200
The product is specific for Humanized VHH antibodies and Camelid (Llama Alpaca and Camel) heavy chain antibodies (IgG2and3) but does not recognize the camelid conventional antibody (IgG1).