Learn More
Genscript Corporation MonoRab™ Rabbit Anti-Humanized VHH Antibody, mAb
Supplier:  Genscript Corporation A02165200
Encompass
A camelid single-domain antibody (sdAb, called Nanobody by Ablynx) is a peptide chain about 110 amino acids long comprising one variable domain of a heavy-chain antibody (VHH fragment). Like a whole antibody, a single-domain antibody is able to bind selectively to a specific antigen. Such antibodies allow a broad range of applications in biotechnical as well as therapeutic fields due to their small size, simple production, and high affinity. However, their sequences may increase the risk of immunogenicity and cause anti-drug antibody (ADA) development in humans, and thus, sdAbs are routinely humanized during development.
Catalog No. NC2132538