Learn More
Genscript Corporation MonoRab Rabbit Anti-scFv Cock
Supplier: Genscript Corporation A02284200
This product is specific for scFvs in different species (humanized mouse) and different orders (VH-linker-VL VL-linker-VH) It can also recognize various forms of scFv-based bispecific antibodies such as DART BITE TandAbs IgG(H)-scFv2 scFv-Fab-Fc etc