Learn More
Genscript Corporation MonoRab Rabbit Anti-scFv Cock
Catalog No. 5000058374
Encompass
This product is specific for scFvs in different species (humanized mouse) and different orders (VH-linker-VL VL-linker-VH) It can also recognize various forms of scFv-based bispecific antibodies such as DART BITE TandAbs IgG(H)-scFv2 scFv-Fab-Fc etc
Catalog No. 50-000-58374 Supplier Genscript Corporation Supplier No. A02286200
This product is specific for scFvs in different species (humanized mouse) and different orders (VH-linker-VL VL-linker-VH) It can also recognize various forms of scFv-based bispecific antibodies such as DART BITE TandAbs IgG(H)-scFv2 scFv-Fab-Fc etc