Learn More
Genscript Corporation MonoRab Rabbit Anti-scFv Cock
Supplier: Genscript Corporation A02304200
This product is specific for scFvs in various species (humanized mouse) various orders (VH-linker-VL VL-linker-VH) and various forms of scFv-based bispecific antibodies(DART BiTE TandAbs IgG(H)-scFv2 scFv-Fab-Fc etc) This product exhibits minimal cross-reactivity with human mouse goat and rabbit immunoglobulins