Learn More
Genscript Corporation MonoRab SARS-CoV-2 Spike 417
Catalog No. 5000058955
Encompass
GenScript MonoRab SARS-CoV-2 Spike (417N) Neutralizing Antibody (21F1) mAb Rabbit is specific for 417N site of B 1 351 (SARS-CoV-2 Beta VOC) RBD This product can recognize RBD single mutant (K417N) and RBD triple mutant (K417N E484K and N501Y) but can t recognize the wild type RBD RBD single mutant (N501Y) and RBD single mutant (E484K)
Catalog No. 50-000-58955 Supplier Genscript Corporation Supplier No. A021401
GenScript MonoRab SARS-CoV-2 Spike (417N) Neutralizing Antibody (21F1) mAb Rabbit is specific for 417N site of B 1 351 (SARS-CoV-2 Beta VOC) RBD This product can recognize RBD single mutant (K417N) and RBD triple mutant (K417N E484K and N501Y) but can t recognize the wild type RBD RBD single mutant (N501Y) and RBD single mutant (E484K)