Learn More
Genscript Corporation MonoRabRabbit Anti-scFv Cock
Catalog No. 5000057848
Encompass
This product is specific for scFvs in various species (humanized mouse) various orders (VH-linker-VL VL-linker-VH) and various forms of scFv-based bispecific antibodies(DART BiTE TandAbs IgG(H)-scFv2 scFv-Fab-Fc etc) This product exhibits minimal cross-reactivity with human mouse goat and rabbit immunoglobulins
Catalog No. 50-000-57848 Supplier Genscript Corporation Supplier No. A02303200
This product is specific for scFvs in various species (humanized mouse) various orders (VH-linker-VL VL-linker-VH) and various forms of scFv-based bispecific antibodies(DART BiTE TandAbs IgG(H)-scFv2 scFv-Fab-Fc etc) This product exhibits minimal cross-reactivity with human mouse goat and rabbit immunoglobulins