Learn More
QA Bio Inc Monomix Standard - Monosaccharide Quantitative Standards
Catalog No. 502236576
Encompass
A quantitative mix of six monosaccharides: glucosamine – galactosamine – galactose – mannose – glucose/dextrose – fucose • 10 nmols of each monosaccharide per vial • Form: Dry. Dried by centrifugal evaporation from an aqueous solution.
This item is not returnable. View return policy
Catalog No. 50-223-6576 Supplier QA Bio Inc Supplier No. CMMONOMIX10
A quantitative mix of six monosaccharides: glucosamine – galactosamine – galactose – mannose – glucose/dextrose – fucose • 10 nmols of each monosaccharide per vial • Form: Dry. Dried by centrifugal evaporation from an aqueous solution.