Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monosaccharides Kit, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111004393 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4393 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4393 Supplier MilliporeSigma Supelco Supplier No. 47267
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monosaccharides Kit contains individually packaged standards in the quantities listed. The monosaccharides are prepared, tested, and individually packaged using rigorous manufacturing procedures.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Neat
Quantity 1 Kit
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) D-(-)Arabinose 500 mg; Fructose 500 mg; D-(+)Galactose 500 mg; D-(+)Glucose, mixed anomers (47249) 500 mg; D-(+)Mannose (mixed anomers) 500 mg; D-(-)Ribose 500 mg; D-(+)Xylose 500 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.