Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monosodium 2-sulfoterephthalate | 19089-60-2 | MFCD00059843 | 99.0% | 269.19 | C8H6NaO7S | 5 G
SDP

Catalog No. 5000208783
Encompass_Preferred

Monosodium 2-sulfoterephthalate is a research-use small molecule reported to inhibit glutamate carboxypeptidase II. It is supplied as a high-purity solid with documented solubility and storage recommendations for in vitro and in vivo research.

  • CAS number: 19089-60-2
  • Molecular formula: C8H6NaO7S
  • Molecular weight: 269.19
  • Purity: 99.0%
  • Appearance: white to off-white solid
  • Storage: 4°C, sealed and protected from moisture; in solvent: -80°C (6 months) or -20°C (1 month)
  • Solubility: DMSO 100 mg/mL; in vivo formulations ≥5 mg/mL in specified vehicles
  • Intended use: for research use only, not for human or veterinary use

Catalog No. 50-002-08783 Supplier Medchemexpress LLC Supplier No. HYW0903095G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monosodium 2-sulfoterephthalate is a research-use small molecule reported to inhibit glutamate carboxypeptidase II. It is supplied as a high-purity solid with documented solubility and storage recommendations for in vitro and in vivo research.

  • CAS number: 19089-60-2
  • Molecular formula: C8H6NaO7S
  • Molecular weight: 269.19
  • Purity: 99.0%
  • Appearance: white to off-white solid
  • Storage: 4°C, sealed and protected from moisture; in solvent: -80°C (6 months) or -20°C (1 month)
  • Solubility: DMSO 100 mg/mL; in vivo formulations ≥5 mg/mL in specified vehicles
  • Intended use: for research use only, not for human or veterinary use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.