Learn More
Selleck Chemical LLC L-Glutamic acid monosodium salt S4721-100mg
SDP
Catalog No. 503110725
Encompass_Preferred
L-Glutamic acid monosodium salt is the sodium salt of glutamic acid found naturally in tomatoes cheese and other foods L-Glutamic acid monosodium salt acts as an excitatory transmitter and an agonist at all subtypes of glutamate receptors (metabotropic kainate NMDA and AMPA) (S)-Glutamic acid shows a direct activating effect on the release of DA from dopaminergic terminals
Catalog No. 50-311-0725 Supplier Selleck Chemical LLC Supplier No. S4721100MG
L-Glutamic acid monosodium salt is the sodium salt of glutamic acid found naturally in tomatoes cheese and other foods L-Glutamic acid monosodium salt acts as an excitatory transmitter and an agonist at all subtypes of glutamate receptors (metabotropic kainate NMDA and AMPA) (S)-Glutamic acid shows a direct activating effect on the release of DA from dopaminergic terminals