Learn More
Sigma Aldrich Fine Chemicals Biosciences Monosodium Glutamate Pharm1G
Catalog No. 502904387
Encompass
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Catalog No. 50-290-4387 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PHR26341G
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.