Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monosodium Maleate Trihydrate 98.0+%, TCI America™
SDP

Catalog No. M001625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M001625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M001625G Supplier TCI America Supplier No. M001625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3105-55-3
Molecular Formula C4H2NaO4
Molecular Weight (g/mol) 137.05
MDL Number MFCD00070246
InChI Key VRVKOZSIJXBAJG-ODZAUARKSA-L
Synonym sodium hydrogen maleate, monosodium maleate, maleic acid monosodium salt, unii-eel2016s8m, maleic acid, monosodium salt, ccris 1121, maleic acid sodium salt, sodium z-3-carboxyacrylate, 2-butenedioic acid 2z-, monosodium salt, sodium poly maleate
PubChem CID 91658932
IUPAC Name sodium (2Z)-but-2-enedioate
SMILES [Na+].[O-]C(=O)\C=C/C([O-])=O

Specifications

Color White
Quantity 25 g
Formula Weight 138.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Monosodium Maleate Trihydrate
EINECSNumber (2)-1102
RTECSNumber ON3500000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.