Learn More
AdipoGen Monosodium urate [MSU] (crystals),Chemical. CAS: 1198-77-2.
SDP
Supplier:  AdipoGen AGCR139502002
Encompass_Preferred
Monosodium urate [MSU] (crystals), AdipoGen Life Sciences. Chemical. CAS: 1198-77-2. Formula: C5H3N4O3 . Na. MW: 167.1 . 23.0. NLRP3/NALP3 inflammasome activator. Specially crystallized and tested for biological activity. Stimulates the caspase-1 activating NLRP3/NALP3 inflammasome, resulting in the production of active IL-1beta and IL-18.
Catalog No. 50-464-781