Learn More
AdipoGen Monosodium urate crystals
SDP
Catalog No. 502562166
Encompass_Preferred
Chemical. CAS 1198-77-2. Formula C5H3N4O3 . Na. MW 167.1 . 23.0. NLRP3/NALP3 inflammasome activator. Specially crystallized and tested for biological activity. Stimulates the caspase-1 activating NLRP3/NALP3 inflammasome, resulting in the production of active IL-1beta and IL-18.
Catalog No. 50-256-2166 Supplier AdipoGen Supplier No. AGCR13950M002
Chemical. CAS 1198-77-2. Formula C5H3N4O3 . Na. MW 167.1 . 23.0. NLRP3/NALP3 inflammasome activator. Specially crystallized and tested for biological activity. Stimulates the caspase-1 activating NLRP3/NALP3 inflammasome, resulting in the production of active IL-1beta and IL-18.