Learn More
Abcam Monosodium urate (PBS solution), NLRP3/NALP3 inflammasome activator, 10MG
Catalog No. 502705902
Encompass_Preferred
MW 190.09 Da. NLRP3/NALP3 inflammasome activator. Stimulates caspase-1 activating NLRP3/NALP3 inflammasome, results in active IL-1β and IL-18 production. Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5902 Supplier Abcam Supplier No. AB14430610MG
MW 190.09 Da. NLRP3/NALP3 inflammasome activator. Stimulates caspase-1 activating NLRP3/NALP3 inflammasome, results in active IL-1β and IL-18 production. Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement