Learn More
GL Sciences Inc MonoSpin Reversed Phase Column, C18, S Type, 50/pack
Catalog No. NC1609946
Encompass
MonoSpin is a monolithic SPE column excellent for the small volume sample with an easy and quick operation by centrifuge. Octadecyl functional group. Optimal for drug extraction in biological samples, and desalting & enrichment of peptide samples. Sample volume : up to 800 μL. Eluent volume : 50 μL - 800 μL. Spin Column is used setting with 1.5 mL and 2 mL micro tubes.
Catalog No. NC1609946 Supplier GL Sciences Inc Supplier No. 501021700
MonoSpin is a monolithic SPE column excellent for the small volume sample with an easy and quick operation by centrifuge. Octadecyl functional group. Optimal for drug extraction in biological samples, and desalting & enrichment of peptide samples. Sample volume : up to 800 μL. Eluent volume : 50 μL - 800 μL. Spin Column is used setting with 1.5 mL and 2 mL micro tubes.