Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monostearyl maleate | 2424-62-6 | MFCD08274815 | 98.0% | 368.55 | C22H40O4 | 10 G
SDP

Catalog No. 5000282695
Encompass_Preferred

Monostearyl maleate (octadecyl fumarate) is an ester reagent used in laboratory research, typically for chemical synthesis, formulation studies, and materials research. It is supplied as a white to off-white solid and is accompanied by manufacturer documentation (datasheet, SDS, COA) for safety and quality verification. This material is intended for research use only and not for clinical or human applications.

  • Purity 98.0%.
  • CAS number 2424-62-6.
  • Molecular formula C22H40O4; molecular weight 368.55.
  • White to off-white solid physical form.
  • Supplied with datasheet, SDS, and COA for verification.
  • Available in small pack sizes suitable for research, including 10 G.
  • Intended for laboratory research use only, not for clinical or human use.

Catalog No. 50-002-82695 Supplier Medchemexpress LLC Supplier No. HYW14048010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monostearyl maleate (octadecyl fumarate) is an ester reagent used in laboratory research, typically for chemical synthesis, formulation studies, and materials research. It is supplied as a white to off-white solid and is accompanied by manufacturer documentation (datasheet, SDS, COA) for safety and quality verification. This material is intended for research use only and not for clinical or human applications.

  • Purity 98.0%.
  • CAS number 2424-62-6.
  • Molecular formula C22H40O4; molecular weight 368.55.
  • White to off-white solid physical form.
  • Supplied with datasheet, SDS, and COA for verification.
  • Available in small pack sizes suitable for research, including 10 G.
  • Intended for laboratory research use only, not for clinical or human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.