Learn More
AdipoGen Monosultap
SDP
Catalog No. 502560485
Encompass_Preferred
Chemical. CAS 29547-00-0. Formula C5H12NNaO6S4. MW 333.4. Synthetic. Nereistoxin analog insecticide. Insect nicotinic acetylcholine receptors nAChRs channel blocker. Compound can be used as analytical reference material.
Catalog No. 50-256-0485 Supplier AdipoGen Supplier No. CDXM0164M100
Chemical. CAS 29547-00-0. Formula C5H12NNaO6S4. MW 333.4. Synthetic. Nereistoxin analog insecticide. Insect nicotinic acetylcholine receptors nAChRs channel blocker. Compound can be used as analytical reference material.