Learn More
AdipoGen Monosultap
SDP
Supplier:  AdipoGen CDXM0164M250
Encompass_Preferred
Chemical. CAS 29547-00-0. Formula C5H12NNaO6S4. MW 333.4. Synthetic. Nereistoxin analog insecticide. Insect nicotinic acetylcholine receptors nAChRs channel blocker. Compound can be used as analytical reference material.
Catalog No. 50-256-0484