Learn More
Med Vet International Monosyn, DS19-4/0, 45CM, JG22504, 12 Count
Catalog No. 5000581191
Encompass_Preferred
Absorbable monofilament suture made of glyconate.Considered a midterm suture in regard to absorption as it retains 50 percent of tensile strength up to 14 days.Exceptional knot security and knot tensile strength.Smooth monofilament for ease of passage through tissue.Braided suture "feel"Absorbed via hydrolysisViolet color for easy identification
Catalog No. 50-005-81191 Supplier Med Vet International Supplier No. JORG22504
Absorbable monofilament suture made of glyconate.Considered a midterm suture in regard to absorption as it retains 50 percent of tensile strength up to 14 days.Exceptional knot security and knot tensile strength.Smooth monofilament for ease of passage through tissue.Braided suture "feel"Absorbed via hydrolysisViolet color for easy identification