Learn More
Med Vet International Monosyn Violet HS26-2/0 90cm, JG22299, 12 Count
Catalog No. 5000581349
Encompass_Preferred
Absorbable monofilament suture made of glyconate.Considered a midterm suture in regard to absorption as it retains 50 percent of tensile strength up to 14 days.Exceptional knot security and knot tensile strength.Smooth monofilament for ease of passage through tissue.Braided suture "feel"Absorbed via hydrolysisViolet color for easy identification
Catalog No. 50-005-81349 Supplier Med Vet International Supplier No. JORG22299
Absorbable monofilament suture made of glyconate.Considered a midterm suture in regard to absorption as it retains 50 percent of tensile strength up to 14 days.Exceptional knot security and knot tensile strength.Smooth monofilament for ease of passage through tissue.Braided suture "feel"Absorbed via hydrolysisViolet color for easy identification