Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monothioglycerol, 95%, Spectrum™ Chemical
SDP

Catalog No. 18604690 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
50 lb.
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-690 50 lb. Amber Glass Bottle
18-604-688 100 mL Amber Glass Bottle
18-604-689 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-604-690 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M117750LBBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

M1177, 96-27-5, C3H8O2S

Spectrum™ Chemical Monothioglycerol , also known as 3-Mercaptopropane-1,2-diol and thioglycerol, is often used as a matrix in liquid secondary ion mass spectrometry and fast atom bombardment mass spectrometry. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 96-27-5
Molecular Formula C3H8O2S
Molecular Weight (g/mol) 108.16
MDL Number MFCD00004879
InChI Key PJUIMOJAAPLTRJ-UHFFFAOYNA-N
IUPAC Name 3-sulfanylpropane-1,2-diol
SMILES OCC(O)CS

Specifications

CAS Max % 1
Boiling Point 118°C (244.4°F)
Packaging Amber Glass Bottle
Quantity 50 lb.
Percent Purity 95%
Grade Reagent
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Monothioglycerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.