Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monothioglycerol, NF, 97-101%, Spectrum™ Chemical
SDP

Catalog No. 18604691 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-691 25 mL Amber Glass Bottle
18-604-693 500 mL Amber Glass Bottle
18-604-692 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-604-691 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M408025ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

M4080, 96-27-5, C3H8O2S

Spectrum™ Chemical Monothioglycerol, NF

Chemical Identifiers

CAS 96-27-5
Molecular Formula C3H8O2S
Molecular Weight (g/mol) 108.16
MDL Number MFCD00004879
InChI Key PJUIMOJAAPLTRJ-UHFFFAOYNA-N
IUPAC Name 3-sulfanylpropane-1,2-diol
SMILES OCC(O)CS

Specifications

CAS Max % 1
pH 3.5 to 7.0
Boiling Point 118°C (244.2°F)
Ignition Residue 0.001
Packaging Amber Glass Bottle
Refractive Index 1.521-1.526
Quantity 25 mL
Specific Gravity 1.241-1.250
Percent Purity 97 to 101%
Grade NF
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Monothioglycerol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.