Learn More
Medchemexpress LLC Monotropein | 5945-50-6 | MFCD00238650 | 99.1% | 10 MG
SDP
Catalog No. 5000295953
Encompass_Preferred
Monotropein is an iridoid glycoside isolated from Morinda officinalis, supplied as an analytical standard for research and analytical applications. It is used as a reference standard in qualitative and quantitative assays and for method development in HPLC, GC, and mass spectrometry, and is commonly applied in studies of inflammation and tissue-protective effects.
  • High purity analytical standard for assay calibration and method development.
  • Suitable for HPLC, GC, and MS applications.
  • Offered in small, laboratory-friendly solid pack sizes (including 10 mg).
  • Characterized by molecular formula C16H22O11 and CAS 5945-50-6 for unambiguous identification.
Catalog No. 50-002-95953 Supplier Medchemexpress LLC Supplier No. HYN064810MG
Monotropein is an iridoid glycoside isolated from Morinda officinalis, supplied as an analytical standard for research and analytical applications. It is used as a reference standard in qualitative and quantitative assays and for method development in HPLC, GC, and mass spectrometry, and is commonly applied in studies of inflammation and tissue-protective effects.
  • High purity analytical standard for assay calibration and method development.
  • Suitable for HPLC, GC, and MS applications.
  • Offered in small, laboratory-friendly solid pack sizes (including 10 mg).
  • Characterized by molecular formula C16H22O11 and CAS 5945-50-6 for unambiguous identification.