Learn More
Ethos Biosciences Monsels Solution 2oz
Catalog No. 5000819200
Encompass_Preferred
Ferric Subsulfate Solution Monsel s Solution is a dark reddish-brown viscous aqueous liquid with a characterisitic metal odor This is a ready-to-use solution May thicken or crystallize at low temperatures For professional and laboratory use only
Catalog No. 50-008-19200 Supplier Ethos Biosciences Supplier No. 339002
Ferric Subsulfate Solution Monsel s Solution is a dark reddish-brown viscous aqueous liquid with a characterisitic metal odor This is a ready-to-use solution May thicken or crystallize at low temperatures For professional and laboratory use only