Learn More
Cayman Chemical Monosialoganglioside Mixture, 1 mL
SDP
Catalog No. 502989442
Encompass_Preferred
A mixture of monosialogangliosides; intended for use as a reference standard for the detection of these monosialogangliosides in various sample types by TLC or LC-MS
Catalog No. 50-298-9442 Supplier Cayman Chemical Supplier No. 293591 ML
A mixture of monosialogangliosides; intended for use as a reference standard for the detection of these monosialogangliosides in various sample types by TLC or LC-MS