Learn More
Cayman Chemical Monensin (sodium salt), Purity: ≥98%, CAS: 22373-78-0, 500 mg
SDP
Catalog No. 502971376
Encompass_Preferred
An ionophorous antibiotic that preferentially forms complexes with monovalent cations for lipid-soluble transport across membrane bilayers; active against Gram-positive bacteria such as Micrococcus, Bacillus, and Staphylococcus (MICs = 1-12.5 µg/ml), of P. falciparum and Coccicdium protozoa, and certain viruses
Catalog No. 50-297-1376 Supplier Cayman Chemical Supplier No. 16488500 MG
An ionophorous antibiotic that preferentially forms complexes with monovalent cations for lipid-soluble transport across membrane bilayers; active against Gram-positive bacteria such as Micrococcus, Bacillus, and Staphylococcus (MICs = 1-12.5 µg/ml), of P. falciparum and Coccicdium protozoa, and certain viruses