Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHECKERS™ Monster General Purpose Wheel Chock
GREENER_CHOICE

Catalog No. 19085969 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-085-969 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-085-969 Supplier CHECKERS™ Supplier No. UC1700
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Checkers™ Monster General Purpose Wheel Chock are for use with tires with up to a 35 in. outside diameter

  • Rated for vehicles with a gross operating weight up to 30,000 lbs
  • Weather-resistant, lightweight polyurethane construction
  • Resistant to oils, fuels, and solvents
  • Recessed carrying handle
  • Mounting hole for ropes or chains
  • High visibility orange color

Specifications

Color Orange
Packaging Quantity 1 Ea.
Weight (English) 2 lb.
For Use With (Equipment) Tires with up to a 35 in. outside diameter
Material Polyurethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.