Learn More
SEPPIC INC Montanide ISA-720 VG ST, water-in-oil emulsion adjuvant, 10 mL package
Catalog No. NC0685296
Encompass
Montanide ISA-720 VG ST is a water-in-oil emulsion adjuvant mainly composed of mineral oil, emulsifiers, and water, designed to enhance immune responses in vaccines by forming stable emulsions. It is reported to be discontinued, with no current official confirmation of availability from the supplier. Despite listings from distributors, the product does not feature in the official catalog of the supplier, leaving its current status unclear. Historically, it was known for applications in therapeutic vaccines and was explored in clinical trials for various diseases.
  • Composed of mineral oil, emulsifiers, and water
  • Designed to enhance immune responses in vaccines
  • Facilitates slow release of active ingredients
  • Explored for use in human and veterinary vaccine development
  • Noted for reduced inflammatory responses compared to traditional adjuvants
Catalog No. NC0685296 Supplier SEPPIC INC Supplier No. 36059V10PK
Montanide ISA-720 VG ST is a water-in-oil emulsion adjuvant mainly composed of mineral oil, emulsifiers, and water, designed to enhance immune responses in vaccines by forming stable emulsions. It is reported to be discontinued, with no current official confirmation of availability from the supplier. Despite listings from distributors, the product does not feature in the official catalog of the supplier, leaving its current status unclear. Historically, it was known for applications in therapeutic vaccines and was explored in clinical trials for various diseases.
  • Composed of mineral oil, emulsifiers, and water
  • Designed to enhance immune responses in vaccines
  • Facilitates slow release of active ingredients
  • Explored for use in human and veterinary vaccine development
  • Noted for reduced inflammatory responses compared to traditional adjuvants