Learn More
Cayman Chemical MontbretIn A 10mg
SDP
Catalog No. 502990583
Encompass_Preferred
An acylated flavonol glycoside with antidiabetic activity; inhibits human pancreatic α-amylase and B. fibrisolvens amylase B (Kis = 8.1 and 1,600 nM, respectively); reduces blood glucose levels of Zucker diabetic fatty rats at 7.4 mg/kg
Catalog No. 50-299-0583 Supplier Cayman Chemical Supplier No. 3065110 MG
An acylated flavonol glycoside with antidiabetic activity; inhibits human pancreatic α-amylase and B. fibrisolvens amylase B (Kis = 8.1 and 1,600 nM, respectively); reduces blood glucose levels of Zucker diabetic fatty rats at 7.4 mg/kg