Learn More
Cayman Chemical Montelukast, Purity: ≥98%, CAS: 158966-92-8, 50 mg
SDP
Catalog No. 502913017
Encompass_Preferred
A CysLT1 receptor antagonist (IC50 = 4.9 nM in HEK293 cell membranes expressing the human receptor); selective for CysLT1 over CysLT2 receptors (IC50 = >10,000 nM in COS-7 cell membranes expressing the human receptor); inhibits LTD4-induced bronchoconstriction in anesthetized guinea pigs (ED50 = 69 nmol/kg, p.o.); inhibits ovalbumin-induced airway hyperresponsiveness and increases in the number of total cells and eosinophils in BALF in a mouse model of allergic asthma at 3 and 10 mg/kg
Catalog No. 50-291-3017 Supplier Cayman Chemical Supplier No. 3577950 MG
A CysLT1 receptor antagonist (IC50 = 4.9 nM in HEK293 cell membranes expressing the human receptor); selective for CysLT1 over CysLT2 receptors (IC50 = >10,000 nM in COS-7 cell membranes expressing the human receptor); inhibits LTD4-induced bronchoconstriction in anesthetized guinea pigs (ED50 = 69 nmol/kg, p.o.); inhibits ovalbumin-induced airway hyperresponsiveness and increases in the number of total cells and eosinophils in BALF in a mouse model of allergic asthma at 3 and 10 mg/kg