Learn More
Cayman Chemical Montelukast sodium salt
SDP
Catalog No. 502890118
Encompass_Preferred
A CysLT1 receptor antagonist (IC50 = 4.9 nM in HEK293 cell membranes expressing the human receptor); selective for CysLT1 over CysLT2 receptors (IC50 = >10,000 nM in COS-7 cell membranes expressing the human receptor); inhibits LTD4-induced bronchoconstriction in anesthetized guinea pigs (ED50 = 69 nmol/kg, p.o.); inhibits ovalbumin-induced airway hyperresponsiveness and increases in the number of total cells and eosinophils in BALF in a mouse model of allergic asthma at 3 and 10 mg/kg
Catalog No. 50-289-0118 Supplier Cayman Chemical Supplier No. 10008318100 MG
A CysLT1 receptor antagonist (IC50 = 4.9 nM in HEK293 cell membranes expressing the human receptor); selective for CysLT1 over CysLT2 receptors (IC50 = >10,000 nM in COS-7 cell membranes expressing the human receptor); inhibits LTD4-induced bronchoconstriction in anesthetized guinea pigs (ED50 = 69 nmol/kg, p.o.); inhibits ovalbumin-induced airway hyperresponsiveness and increases in the number of total cells and eosinophils in BALF in a mouse model of allergic asthma at 3 and 10 mg/kg