Learn More
Medicus Health Monster Test Tube Racks, 10-13Mm, Holds 216 Test Tubes, Blue
SDP
Catalog No. NC1765228
Encompass
∙ Monster Racks hold a monster load of tubes and are now available in colors! ∙Choose from Blue or White ∙Monster Racks are perfect for organizing and storing specimen tubes in any size Laboratory∙Racks have identifying alpha-numeric for quick location and retrieval of tubes∙Holds 216 Tubes from 10-13mm in diameter∙Tubes are spaced far enough apart to make finding repeats, put always and send outs a snap∙Made of autoclavable polypropylene∙Overall Dimensions: 13.25"L x 9.25"W x 3.5"H
This item is not returnable. View return policy
Catalog No. NC1765228 Supplier Medicus Health Supplier No. 5291M4
∙ Monster Racks hold a monster load of tubes and are now available in colors! ∙Choose from Blue or White ∙Monster Racks are perfect for organizing and storing specimen tubes in any size Laboratory∙Racks have identifying alpha-numeric for quick location and retrieval of tubes∙Holds 216 Tubes from 10-13mm in diameter∙Tubes are spaced far enough apart to make finding repeats, put always and send outs a snap∙Made of autoclavable polypropylene∙Overall Dimensions: 13.25"L x 9.25"W x 3.5"H