Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Montmorillonite

Catalog No. 698661KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-866-1KG 1 kg
69-866-100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-866-1KG Supplier Sigma Aldrich Supplier No. 698661KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Montmorillonite is a layered clay mineral (2:1 phyllosilicate) that belongs to a class of the smectite group. It is a naturally abundant material and is chemically modified by the cation-exchange method for the preparation of polymer nanocomposites.Montmorillonite has been used as:A nanofiller to improve the mechanical properties of poly(vinyl alcohol) (PVA) film for potential usage in packaging applications.A catalyst in the conversion of polar and non-polar algae oil lipids to biodiesel.It can also be used:As a nanofiller in the preparation of biopolymer based nanocomposites.In the preparation of Fe3+-montmorillonite, a solid acid catalyst for the synthesis of xanthenedione derivatives.Solid Lewis acid catalyst

Specifications

CAS 1318-93-0
Quantity 1 kg
MDL Number MFCD00132796
Synonym K-Catalyst
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.