Learn More
Medchemexpress LLC Montmorillonite | 1318-93-0 | ≥98.0% | 1 KG
SDP
Catalog No. 5000410322
Encompass_Preferred
Montmorillonite is composed of two layers of co-top connected silicon oxygen tetrahedral sheet and one layer of co-edge connected aluminum (magnesium) oxygen (hydroxide) octahedral sheet, forming a 2:1 structure containing water of crystal. It can be utilized for large-scale industrial decontamination, effectively removing harmful ions from sewage. Additionally, due to its exchangeable hydrated cations that can adsorb bacteria, montmorillonite can serve as a carrier for ionic antibacterial agents.
  • Effectively removes harmful ions from sewage
  • Can serve as a carrier for ionic antibacterial agents
Catalog No. 50-004-10322 Supplier Medchemexpress LLC Supplier No. HYB21431KG
Montmorillonite is composed of two layers of co-top connected silicon oxygen tetrahedral sheet and one layer of co-edge connected aluminum (magnesium) oxygen (hydroxide) octahedral sheet, forming a 2:1 structure containing water of crystal. It can be utilized for large-scale industrial decontamination, effectively removing harmful ions from sewage. Additionally, due to its exchangeable hydrated cations that can adsorb bacteria, montmorillonite can serve as a carrier for ionic antibacterial agents.
  • Effectively removes harmful ions from sewage
  • Can serve as a carrier for ionic antibacterial agents