Learn More
Medchemexpress LLC Montmorillonite | 1318-93-0 | ≥98.0% | 500 G
SDP
Catalog No. 5000411315
Encompass_Preferred
Montmorillonite is composed of two layers of co-top connected silicon oxygen tetrahedral sheet and one layer of co-edge connected aluminum (magnesium) oxygen (hydroxide) octahedral sheet, forming a 2:1 structure containing water of crystal.
  • Can be used for large-scale industrial decontamination.
  • Effectively removes harmful ions in sewage.
  • Can be used as a carrier of ionic antibacterial agents due to its exchangeable hydrated cations, which can adsorb bacteria.
Catalog No. 50-004-11315 Supplier Medchemexpress LLC Supplier No. HYB2143500G
Montmorillonite is composed of two layers of co-top connected silicon oxygen tetrahedral sheet and one layer of co-edge connected aluminum (magnesium) oxygen (hydroxide) octahedral sheet, forming a 2:1 structure containing water of crystal.
  • Can be used for large-scale industrial decontamination.
  • Effectively removes harmful ions in sewage.
  • Can be used as a carrier of ionic antibacterial agents due to its exchangeable hydrated cations, which can adsorb bacteria.