Learn More
MOORE INDUSTRIES INTERNATIONAL Moore Industries PC-Programmable Isolated Temperature Transmitter, 4-20mA, 10-30DC, Hockey-puck Housing
Catalog No. NC3989623
Encompass
The PC-programmable isolated temperature transmitter combines smart digital technology with advanced analog operation for superior reliability, accuracy, and ease of use. It can be programmed to accept direct inputs from various sources and converts the input to a highly accurate 4-20mA output.
  • Fast measurement cycle with an output update up to eight times per second.
  • Enhanced free configuration software.
  • Programmable input for RTD, T/C, Ohms, mV or potentiometer.
  • Output: 4-20mA.
  • Power: 10-30DC, 2-wire, output loop powered.
  • Hockey-puck housing for mounting in standard connection heads.
  • Fully isolated.
  • CSA approved IS and NI hazardous area certifications.
  • RoHS compliant.
  • PC configurable via USB cable.
Catalog No. NC3989623 Supplier MOORE INDUSTRIES INTERNATIONAL Supplier No. TRYPRG420MA1030DCISCHPPDN
The PC-programmable isolated temperature transmitter combines smart digital technology with advanced analog operation for superior reliability, accuracy, and ease of use. It can be programmed to accept direct inputs from various sources and converts the input to a highly accurate 4-20mA output.
  • Fast measurement cycle with an output update up to eight times per second.
  • Enhanced free configuration software.
  • Programmable input for RTD, T/C, Ohms, mV or potentiometer.
  • Output: 4-20mA.
  • Power: 10-30DC, 2-wire, output loop powered.
  • Hockey-puck housing for mounting in standard connection heads.
  • Fully isolated.
  • CSA approved IS and NI hazardous area certifications.
  • RoHS compliant.
  • PC configurable via USB cable.