Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moore Medical MooreBrand™ Ointment

Catalog No. 19898753
Change view
Click to view available options
Volume (Metric):
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-898-753 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-898-753 Supplier Moore Medical Supplier No. 82466

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Specifications

Product Type Triple Antibiotic Ointment
Active Ingredients Bacitracin (400U), Neomycin Sulfate (5 mg), Polymyxin B Sulfate (5000U)
For Use With (Application) Helps to prevent infection in minor cuts, scrapes and burns
Quantity 144 Pk.
Volume (English) 0.035 oz.
Volume (Metric) 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.