Learn More
World Precision Instrument Moorfield Forceps, 11 cm, Straight, 0.9 mm Serrated tips
SDP
Catalog No. 502538261
Encompass
Moorfield Forceps, measuring 11 cm (4.3") in length, are specialized ophthalmic forceps used in eye surgery, particularly for procedures involving the anterior segment of the eye. These forceps are available in two materials: stainless steel or black titanium-coated stainless steel. The straight design of Moorfield forceps allows for precise grasping and manipulation of delicate tissues during surgeries such as cataract surgery, corneal transplantation, and other anterior segment procedures.
Catalog No. 50-253-8261 Supplier World Precision Instrument Supplier No. WPF5055
Moorfield Forceps, measuring 11 cm (4.3") in length, are specialized ophthalmic forceps used in eye surgery, particularly for procedures involving the anterior segment of the eye. These forceps are available in two materials: stainless steel or black titanium-coated stainless steel. The straight design of Moorfield forceps allows for precise grasping and manipulation of delicate tissues during surgeries such as cataract surgery, corneal transplantation, and other anterior segment procedures.