Learn More
Andwin Scientific MOPHEAD CLEANRM BLUE 14IN CS12
SDP
Catalog No. NC0092737
Encompass_Preferred
The Roll-O-Matic Sponge Refills are ideal for general use within all controlled environments. This product is made to absorb spills, clean particles, and apply disinfectant. It is autoclavable and resistant to gamma and ETO sterilization. The refill
Catalog No. NC0092737 Supplier Andwin Scientific Supplier No. 89032176
The Roll-O-Matic Sponge Refills are ideal for general use within all controlled environments. This product is made to absorb spills, clean particles, and apply disinfectant. It is autoclavable and resistant to gamma and ETO sterilization. The refill