Learn More
TARGETMOL CHEMICALS INC MOPIDAMOL 5MG
SDP
Catalog No. 502580738
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Mopidamol (RA 233) is a phosphodiesterase inhibitor a dipyridamole derivative with anticancer activity that prevents retinal vascular defects in experimental diabetes. purity: 99%
Catalog No. 50-258-0738 Supplier TARGETMOL CHEMICALS INC Supplier No. T258295MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Mopidamol (RA 233) is a phosphodiesterase inhibitor a dipyridamole derivative with anticancer activity that prevents retinal vascular defects in experimental diabetes. purity: 99%