Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mopivabil | 2988562-84-9 | >98.0% | C14H20O3 | 10MG
SDP

Catalog No. 5000258696
Encompass_Preferred

Mopivabil is a research reagent reported to act as an angiotensin II receptor antagonist, intended for in vitro pharmacology and biochemical studies. The compound is supplied as a high-purity solid and is commonly prepared in DMSO for assay use. Typical analytical data include formula C14H20O3, molecular weight 236.31, and CAS 2988562-84-9.

  • High purity suitable for research (>98%).
  • Chemical formula C14H20O3 and molecular weight 236.31.
  • Soluble in DMSO for convenient assay preparation.
  • Available as solid and DMSO solution formats for flexibility.
  • Suitable for in vitro studies of angiotensin II receptor signaling.
  • Storage stability in solvent: -80°C up to 6 months, -20°C up to 1 month.

Catalog No. 50-002-58696 Supplier Medchemexpress LLC Supplier No. HY14561110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mopivabil is a research reagent reported to act as an angiotensin II receptor antagonist, intended for in vitro pharmacology and biochemical studies. The compound is supplied as a high-purity solid and is commonly prepared in DMSO for assay use. Typical analytical data include formula C14H20O3, molecular weight 236.31, and CAS 2988562-84-9.

  • High purity suitable for research (>98%).
  • Chemical formula C14H20O3 and molecular weight 236.31.
  • Soluble in DMSO for convenient assay preparation.
  • Available as solid and DMSO solution formats for flexibility.
  • Suitable for in vitro studies of angiotensin II receptor signaling.
  • Storage stability in solvent: -80°C up to 6 months, -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.