Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mopivabil | 2988562-84-9 | 99.7% | C14H20O3 | 50 MG
SDP

Catalog No. 5000355287
Encompass_Preferred

Mopivabil is an angiotensin II receptor antagonist.

  • Molecular formula: C₁₄H₂₀O₃
  • Molecular weight: 236.31
  • Appearance: Colorless to light yellow oil
  • Purity (LCMS): 99.66%
  • Storage (pure form): -20°C for 3 years; 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month

Catalog No. 50-003-55287 Supplier Medchemexpress LLC Supplier No. HY14561150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mopivabil is an angiotensin II receptor antagonist.

  • Molecular formula: C₁₄H₂₀O₃
  • Molecular weight: 236.31
  • Appearance: Colorless to light yellow oil
  • Purity (LCMS): 99.66%
  • Storage (pure form): -20°C for 3 years; 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.