Learn More
MSC PortionPac Floor Cleaner, pH Neutral, Biodegradable, 2 gal
Catalog No. 502866971
Encompass
MopPacLite pH Neutral Floor Cleaner gently cleans all hard surface flooring without leaving behind any dulling residue. Its biodegradable formulation meets Green Seal Standard GS-37 for effective performance and environmental considerations.
  • Gently cleans all hard surface flooring
  • Leaves no dulling residue
  • Biodegradable formulation
  • Meets Green Seal Standard GS-37 based on effective performance, concentrated volume, minimized/recycled packaging, and protective limits on VOCs and human & environmental toxicity
Catalog No. 50-286-6971 Supplier MSC Supplier No. 38689170
MopPacLite pH Neutral Floor Cleaner gently cleans all hard surface flooring without leaving behind any dulling residue. Its biodegradable formulation meets Green Seal Standard GS-37 for effective performance and environmental considerations.
  • Gently cleans all hard surface flooring
  • Leaves no dulling residue
  • Biodegradable formulation
  • Meets Green Seal Standard GS-37 based on effective performance, concentrated volume, minimized/recycled packaging, and protective limits on VOCs and human & environmental toxicity